Woulff bottles
Katalogové číslo 519-4500
Dodavatel: Sartorius
Specifikace
- Kapacita:500 ml
- Hrdlo:with two side necks GL 18, center neck GL 25, complete with screw caps
- Bal.:1
Specifikace
O tomto produktu
Made of borosilicate glass.
- Side necks with screw caps and inlet/outlet glass tubes
- Centre neck with screw cap and stopcock
- Used between suction flask and vacuum source
- Very good chemical and temperature resistance
With three necks. For protection of vacuum pump.