Orders LinkContactUsLinkComponent
Detergent, Diversey Attack Plus E9e
Detergent, Diversey Attack Plus E9e
01, Diversey Attack Plus E9e, detergent, multi-purpose, 2×5 l containers/case
Katalogové číslo DIVY7516886
Dodavatel:   Diversey
Detergent, Diversey Attack Plus E9e
Katalogové číslo DIVY7516886
Dodavatel:  Diversey
Detergent, Diversey Attack Plus E9e
01, Diversey Attack Plus E9e, detergent, multi-purpose, 2×5 l containers/case
Přejít na:
Price
2 990,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Clear yellow, heavy-duty cleaner containing a mixture of organic solvents and compounds.
  • Good emulsifying formula
  • Removes grease and dirt
  • Medium foaming alkaline formula, pH of neat solution: 12
Specifikace
  • Popis:
    Diversey Attack Plus E9e, detergent, multi-purpose
  • Sekvence:
    01
  • Balené:
    2×5 l containers/case