Disinfectant, Phagospray DM
Dodavatel: PHAGOGENE
Certifikáty
O tomto produktu
Disinfectant based on aqueous alcohol for floors and large surfaces.
- Broad spectrum disinfectant for reducing the risk of contamination
- Contains ethanol (~23%), didecyldimethylammonium chloride, and alkylamine
- Rapid cleaning and disinfection with a simple 'spray and wipe'
- Grapefruit fragrance
Certifications: Bactericide EN 1040, EN 1276, EN 13727, EN 13697; Mycobactericide EN 14348; Fungicide EN 1275, EN 13624, EN13697; Virucide active against Rotavirus and Influenze A/H1N1 and also active against HIV and HBV.
Caution: Use biocides safely. Always read the label and product information before use.
Související informace
Frequently Bought Together