Surface disinfectant, mikrozid® AF liquid
SCUE109160
- Popis:mikrozid® AF liquid, rapid disinfectant,
- Sekvence:01
- Balené:250 ml spray bottle
- Bal.:10
Aldehyde-free rapid disinfectant with broad biocidal activity, used for medical devices.
- Dries rapidly without leaving smears
- Good wetting properties
- Ready to use.