Denatured Ethanol, filtered, for cleanroom use, Klercide 70 | 30 Denatured Ethanol
Catalog # 141-1098
Supplier: ECOLAB CONTAMINATION CONTROL
Specifications
- Popis:Klercide 70 | 30 denatured ethanol
- Balené:5,0 litre container
- Bal.:4
Specifications
About this item
70% denatured ethanol, 30% deionised water solution, 0,2 µm filtered.
- SteriShield Delivery System, which throughout use preserves the integrity of the contents
- Suitable for grade C/D cleanrooms
Certifications: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697
Packaging: Supplied cleanroom processed and double bagged.
Caution: Use biocides safely. Always read the label and product information before use.