Safety Container
Catalog # SEMA3651
Supplier: SEMADENI
Specifications
- Určeno pro:Test tube racks or small vessels with bacteriological or hazardous substances
- Š×H×V vnějš.:330×175×180 mm
- Barva:Bezbarvá
- Bal.:1
- Š×H×V vnitř.:300×125×160 mm
Specifications
About this item
Safety container made of PC, with hermetically sealed lid and stainless steel mounting bracket.
- Robust design
- Avoid direct contact with solvents, strong acids, and alkaline solutions
Ideal for the safe storage and transport of test tube racks and small vessels with bacteriological and hazardous substance.