Orders LinkContactUsLinkComponent
Cover spectacles, Overlight
02, Size L, anti-UV (2C-1.2), anti-fog, anti-scratch, Průhledná, Clear PC
Catalog # BOLEOVLITLPSI
Supplier:   Bollé Safety
Cover spectacles, Overlight
Catalog # BOLEOVLITLPSI
Supplier:  Bollé Safety
Cover spectacles, Overlight
Cover spectacles, Overlight
02, Size L, anti-UV (2C-1.2), anti-fog, anti-scratch, Průhledná, Clear PC
Price
193,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Compact and attractive cover glasses (OTG - over the glasses) spectacles guarantee maximum safety for all.
  • Pivoting temples
  • Available in two sizes
Ideal for wearers of prescription glasses.
Weight: 25 g
 : EN 170; EN 166 FT 1
Specifications
  • Type:
    Size L, anti-UV (2C-1.2), anti-fog, anti-scratch
  • Barva:
    Průhledná
  • Sekvence:
    02
  • Čočky/objektiv:
    Clear PC