Quick release clamp, for SLV® reaction vessels
Catalog # BIBB295-02
Supplier: SciLabware
Specifications
- Určeno pro:100 mm flanges, complete
- Bal.:1
Specifications
About this item
Made of stainless-steel.
- For securing SVL® lids to reaction vessels
- One piece construction
- Can be secured and removed in a single movement with an adjustable locking tab