Orders LinkContactUsLinkComponent
Glucose solution
Glucose solution, 10 mL
Katalogové číslo 201000005.
Dodavatel:   Polyplus-transfection
Glucose solution
Katalogové číslo 201000005.
Číslo CAS:  
Glucose solution
Glucose solution
Glucose solution, 10 mL
Přejít na:
Číslo CAS:  
Price
1 290,00 Kč
Přihlásit se pro dostupnost
1 * 10 mL
Product Details & Documents
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol “New”. For duplicated products with symbol “telephone” please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Specifikace
Specifikace
  • Bal.:
    10 mL
  • Popis:
    Glucose solution