Phenol red, sodium salt
Catalog # P5648-10GR
Supplier: Biowest
CAS Number:
Specifications
- Bal.:10 g
- Popis:Phenol red, sodium salt
Specifications
About this item
Phenol red is used as a pH indicator in cell culture. Its colour exhibits a gradual transition from yellow to red over the pH range 6,8 to 8,2. Above pH 8,2, phenol red turns a bright pink (fuchsia) colour.