Tubing coupling
Catalog # BURK8704-0608
Supplier: BURKLE
Specifications
- Hadice int.Ø:5-7 mm
- Popis:Hose connector, conical, attachable
- Bal.:10
Specifications
About this item
Attachable, conical hose couplings, made of PE, for pressure less operation.
- Straight
- Good chemical resistance
- Good temperature resistance