Biocide, Klercide™ Sporicidal active chlorine unit dose concentrate
Katalogové číslo 141-1204
Dodavatel: ECOLAB CONTAMINATION CONTROL
Specifikace
- Popis:Klercide™ Sporicidal Active Chlorine unit dose concentrate, sterile
- Balené:170 g plastic container
- Bal.:12
Specifikace
O tomto produktu
A unit dose concentrate of Klercide Sporicidal Active Chlorine and an effective sporicide based on active chlorine
- Suitable for class C/D cleanrooms
- Ideal for use with the Klercide Mopping Systems
- Unit dose (in a tub of 17×10 g tablets) for production of 10 litres of solution
- Each tub is filled in a Grade D (ISO Class 8) cleanroom and double bagged, before irradiation at no less than 25 kGy
Certifications: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697; Sporicidal EN 13704.
Caution: Use biocides safely. Always read the label and product information before use.