LED illuminated stand magnifier, visolux+
630-2181
- Zvětšení:3×
- Bal.:1
- Š×H:100×75 mm
This magnifier offers bright and glare-free illumination provided by two SMD LEDs. An integrated pre-selectable orange filter can be used for a warm colour temperature.
- Automatic light off after 30 minutes
- Meniscus/aspheric lens for a larger viewing distance
- Cera-tec® anti-scratch hard coating: Reliability, comfort and relaxed vision
: Supplied with a hard foam case and three AA batteries.