Beard cover, BioClean Beard Snood™
NITRBBS-W
: Nitritex
- Balené:100 pieces per bag; 10 bags per carton
- Barva:Bílá
- Bal.:1.000
Lightweight beard cover made of soft non woven PP. Suitable for use in a variety of environments.
- Soft fabric
- Elasticated openings
- Comfortable headband loop
Width across face: 170 mm ± 5 mm