Magnetic stirring bar retriever, PP
Katalogové číslo DELT19919
Dodavatel: DELTALAB
Specifikace
- Ø:10 mm
- Popis:Stirring bar retriever
- Length:300 mm
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
PP coated stirring bar retriever for the removal of stirrer magnets from vessels of all kinds.
- Magnetic tip for easy removal of magnets