The Q55 sonicator is a compact and effective ultrasonic processor ideal for standard cell disruption and other small volume applications.
- Small footprint - save valuable bench space
- Thumb switch or continuous operation
- Simple controls
A digital programmable sonicator which is effective for standard cell disruption, DNA/RNA shearing, homogenisation and many other applications. The Q125 is ideal for small samples and for customers that do not plan to scale up to larger volumes in the future.
Programmable operation by setting time and amplitude
Pulse mode prevents heat build up in temperature sensitive samples
Digital amplitude/intensity control
Real time energy monitoring
A digital programmable sonicator which is effective for standard cell disruption, DNA/RNA shearing, homogenization and many otherapplications. The Q125 is ideal for small samples and for customers that do not plan to scale up to larger volumes in the future.
- Catalog No:
- 432-0236
- 432-0235
- 432-0232
- 432-0231
- Model:
- Q55
- Q55
- Q125
- Q125
- Frekvence:
- 20 kHz
- 20 kHz
- 20 kHz
- 20 kHz
- Maximální výkon:
- 55 W
- 55 W
- 125 W
- 125 W
- Časovač:
- -
- -
- Up to 10 hours
- Up to 10 hours
- Programovatelné:
- -
- -
- Time and amplitude
- Time and amplitude
- Š×H×V:
- 203×190×146 mm
- 203×190×146 mm
- 203×350×146 mm
- 203×350×146 mm
- Časový interval:
- -
- -
- 1 second to 1 minute
- 1 second to 1 minute