O tomto produktu
Suitable for even the most demanding and sensitive applications, the family of Thermo Scientific™ Barnstead™ GenPure™ water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality.
- UV oxidation reduces organic levels to <5 ppb, and protects against microorganisms (only on systems containing a UV lamp (UV))
- Ultrafiltration removes pyrogens from the feed water
- Feed water monitoring alerts users to fluctuations in feed water quality
- Real-time monitor of the amount of organic materials in the product water shown on the display
- USP-compliant conductivity measurement with temperature compensation can be switched on or off
- Feed water source: Potable tap water, pretreated by reverse osmosis, ion exchange or distillation
- RS-232 interface enables data printing and satisfies GLP guidelines
- User-friendly, illuminated four-line alpha-numeric displays and tilting control panels for optimal view
Special feature of GenPure Pro system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01 to 65 L with an accuracy of <0,5%
Special features of GenPure xCAD Plus system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01 to 65 L with an accuracy of <0,5%
- GenPure xCAD Plus remote dispenser for full control of the system
- Under-the-bench mounting: Ideal solution when bench space is limited
Ideal for molecular biology, microbiology and analytical chemistry applications (e.g. cell and tissue culture, PCR, DNA sequencing, electrophoresis, analytical chemistry, HPLC, CG, CG-MS, ICP-MS, AA, TOC Measurements).
Specifikace
- Catalog No:
- BARN50131952
- BARN50136146
- BARN50131217
- BARN50136151
- BARN50136153
- BARN50136152
- BARN50136171
- BARN50136170
- BARN50136172
- BARN50131922
- BARN50136169
- BARN50131950
- BARN50131948
- Model:
- Barnstead GenPure Pro UV
- Barnstead GenPure xCAD Plus UV/UF – TOC, bench version
- Barnstead GenPure UV/UF
- Barnstead GenPure xCAD Plus UV/UF, bench version
- Barnstead GenPure xCAD Plus UV – TOC, bench version
- Barnstead GenPure xCAD Plus UV, bench version
- Barnstead GenPure xCAD Plus UV – TOC, wall version
- Barnstead GenPure xCAD Plus UV, wall version
- Barnstead GenPure xCAD Plus UV/UF – TOC, wall version
- Barnstead GenPure Pro UV/UF – TOC
- Barnstead GenPure xCAD Plus UV/UF, wall version
- Barnstead GenPure Pro UV/UF
- Barnstead GenPure Pro UV – TOC
- Průtok:
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Up to 2 l/min
- Vodivost:
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- 0,055 µS/cm
- Obsah TOC:
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- 1 - 5 ppb
- Rezistivita při 25 °C:
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- 18,2 MΏ/cm
- Bakterie:
- <0,1 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- <0,01 CFU/ml
- Spotřeba:
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- 0,1 kW
- Š×H×V:
- 330×472×615 mm
- 260×530×725 mm
- 372×337×615 mm
- 260×530×725 mm
- 260×530×725 mm
- 260×530×725 mm
- 260×530×655 mm
- 260×530×655 mm
- 260×530×655 mm
- 330×472×615 mm
- 260×530×655 mm
- 330×472×615 mm
- 330×472×615 mm