Cold trap, Quickfit®
Catalog # 201-3052
Dodavatel: SciLabware
Specifikace
- Popis:Cold trap head, Pyrex® borosilicate glass, socket size 19/26 NS, cone size 34/35 and 19/26 NS, for vacuum systems
- Sekvence:01
- Bal.:1
Specifikace
O tomto produktu
Designed for the vacuum systems which prevents the vapours from the volatile materials from damaging pumps and easy to use.