ContactUsLinkComponent
Air/steam inlet tubes, plain, Quickfit®
Air/steam inlet tubes, plain, Quickfit®
Katalogové číslo 201-3059
Dodavatel:  SciLabware
undefined
Air/steam inlet tubes, plain, Quickfit®
Katalogové číslo 201-3059
Dodavatel:  SciLabware
Specifikace
  • Ø:
    7 mm
  • Délka:
    325 mm
  • Bal.:
    5
Specifikace
O tomto produktu
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air/steam inlet tubes are designed to fit the range of Quickfit® thermometer/inlet adapters having size 13 thread. Glass tube with jet.