Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Indikátory sterilizace
Cleanroom labels, autoclavable
Cleanroom labels, autoclavable
Cleanroom labels, autoclavable
Catalog # 817-0302
Supplier:
Texwipe
Expand image
Cleanroom labels, autoclavable
Catalog # 817-0302
Supplier:
Texwipe
Specifications
Šířka:
50 mm
Popis:
Cleanroom labels, autoclavable
Délka:
50 mm
Bal.:
1 Roll
Specifications
About this item
Product Details
Product Details