Strips of 12 PCR tubes and cap strips
Catalog # DELT4095.4N
Supplier: DELTALAB
Specifications
- Balené:80 strips/case
- Kapacita:0,2 ml
- Barva:Přírodní
- Popis:Strips of 12 tubes and 12 caps
- Bal.:80
Specifications
About this item
Tubes made of PP.
- RNAse, DNAse and pyrogen free
- Excellent chemical resistance
- Excellent chemical resistance
Cap and tube strips allow easy handling and inventory of tubes and caps. It also prevent cross-contamination from tube to tube.