Microcentrifuge tubes
Catalog # DELT200400P
Supplier: DELTALAB
Specifications
- Type:with attached lid, Eppendorf type
- Verze:graduated, conical bottom, non sterile
- Kapacita:1,5 ml
- Bal.:6.000
Specifications
About this item
Manufactured in polypropylene.
- Autoclavable
- Easy opening and closing
- Graduated tube
- It features a frosted area for writing
- Flat cap
- Will withstand centrifugation up to: 12500×g