Orders LinkContactUsLinkComponent
Digital timer
Digital timer
02, Digital timer with alarm and instructions in Spanish, English and French, Šedá, 64×14×61 mm
Katalogové číslo DELT900600
Dodavatel:   DELTALAB
Digital timer
Katalogové číslo DELT900600
Dodavatel:  DELTALAB
Digital timer
02, Digital timer with alarm and instructions in Spanish, English and French, Šedá, 64×14×61 mm
Přejít na:
Price
3 140,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Counts up and down.
  • Capacity: 24 hours
  • Alarm (60 seconds)
  • Incl. clip and small bench stand
Delivery information: Supplied with LR44 battery.
Specifikace
  • Barva:
    Šedá
  • Popis:
    Digital timer with alarm and instructions in Spanish, English and French
  • Sekvence:
    02
  • Š×H×V:
    64×14×61 mm