Orders LinkContactUsLinkComponent
Pasteur pipettes, graduated
Pasteur pipettes, graduated
Supplier:   DELTALAB
Pasteur pipettes, graduated
Supplier:  DELTALAB
Certificates
Pasteur pipettes, graduated
2 Options Available Below
Product Details & Documents
Transparent PE.
  • Sterile pipettes are ethylene oxide sterilised
  • ±10% volumetric tolerance
Documents
Product Family Options
Product Information
  • Sekvence
    Kapacita
    Kapky na ml
    Nasátý objem
    Ø stonku
    Délka
    Stupnice
    Sterilní
    Balené
    Availability
    Price
  • 01
    5,0 ml
    25
    3,1 ml
    5 mm
    154 mm
    0,25 ml
    +
    Peel-pack, 6×500
    Log in for availability
    Log in for availability
    1 * 3.000 KS
    1 * 3.000 KS - 19 170,00 Kč
    19 170,00 Kč
    1 * 3.000 KS
       19 170,00 Kč
  • 02
    7,0 ml
    22
    3,2 ml
    7,8 mm
    154 mm
    0,50 ml
    +
    Individually wrapped, peel-pack, 6×500
    Log in for availability
    Log in for availability
    1 * 3.000 KS
    1 * 3.000 KS - 18 690,00 Kč
    18 690,00 Kč
    1 * 3.000 KS
       18 690,00 Kč