Homogenisers, Mixer B-400
Dodavatel: BUCHI
Certifikáty
O tomto produktu
Mixer B-400 is designed for efficient homogenisation of a wide range of food and feed samples. The process is based on simultaneous grinding and homogenisation of sample materials by means of volumetric reduction using two rotating blades in a beaker. Automatic electronic monitoring of the speed and the torque allows safe and straight forward operation of the grinding process.
Specifikace
- Catalog No:
- 432-1101
- 432-1103
- Model:
- Mixer B-400
- Mixer B-400
- Rozsah rychlosti:
- 9000 min⁻¹
- 9000 min⁻¹
- Třída motoru:
- 1900 W
- 1900 W
- Š×H×V:
- 300×530×510 mm
- 300×530×510 mm
- Hmotnost:
- 26 kg
- 26 kg