Emergency body and eye shower
Catalog # 118-2195
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Specifications
- Popis:Emergency body and eye shower combination, free-standing
- Bal.:1
Specifications
About this item
Free-standing emergency body and eye shower combination made of ABS plastic.
- Eye wash with two spray heads and dust caps
- Eye wash with or without collection basin, or with ABS plastic collection basin and cover
- Tubes powder-coated and galvanized, chrome-plated brass valve
- Pull-rod trigger / 90° rotation