ContactUsLinkComponent
Emergency body and eye shower
Emergency body and eye shower
Catalog # 118-2195
undefined
Emergency body and eye shower
Catalog # 118-2195
Specifications
  • Popis:
    Emergency body and eye shower combination, free-standing
  • Bal.:
    1
Specifications
About this item
Free-standing emergency body and eye shower combination made of ABS plastic.
  • Eye wash with two spray heads and dust caps
  • Eye wash with or without collection basin, or with ABS plastic collection basin and cover
  • Tubes powder-coated and galvanized, chrome-plated brass valve
  • Pull-rod trigger / 90° rotation