ContactUsLinkComponent
Test kit for emergency showers
Test kit for emergency showers
Catalog # 118-2196
undefined
Test kit for emergency showers
Catalog # 118-2196
Specifications
  • Popis:
    Test kit for emergency showers, complete
  • Bal.:
    1
Specifications
About this item
This test kit consists of a collection bucket, a water bag with opening ring and a holding rod. It allows regular checks and maintenance of the emergency shower to be carried out simply and conveniently.