Test kit for emergency showers
Catalog # 118-2196
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Specifications
- Popis:Test kit for emergency showers, complete
- Bal.:1
Specifications
About this item
This test kit consists of a collection bucket, a water bag with opening ring and a holding rod. It allows regular checks and maintenance of the emergency shower to be carried out simply and conveniently.