Deep well microplates, 96-round well
Catalog # 732-2479
Supplier: BIOTIX
Specifications
- Sterilní:+
- Barva:Průhledná
- Popis:Deep well microplate, 96-round well with U-bottom
- Sekvence:01
- Environmentally Preferable:
- Bal.:5
- Well volume:1,2 ml
Specifications
About this item
Medical grade, virgin PP, untreated. With a smooth, round-well geometry, these deep well microplates maximise sample volume while eliminating the 'wicking' effect common with square-well microplates.
- Excellent chemical and temperature resistance
- Tested free from DNase, RNase and endotoxin
Certifications: Conform to ANSI/SBS 1-2004 standards for automated workstations. All plates are BioReady™ certified for purity.