Folding steps
Folding step, non-slip serrated aluminium treads, 2, 2450 mm, 3,9 kg
Katalogové číslo 139-9932
Dodavatel: ZARGES LEICHTBAU
Přejít na:
Product Details & Documents
The Z600 step stool is a quick and flexible solution that can be folded to space-saving size for transport. Stiles made from extruded aluminium for stability and ends with plastic edge finish.
- Minimum space requirement for storage and transport
- Non-slip serrated aluminium treads
Specifikace
- Poč. kroků:2
- Popis:Folding step, non-slip serrated aluminium treads
- Výška:2450 mm
- Hmotnost:3,9 kg