Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
BELAF378760002
- Popis:Bel-Blotter™ replicating tool, 96-well
- Bal.:1
The 96 'open-ended' pipette tips in this replicating tool are compatible with all kinds of plates and tubes.
- Fits plates with flat, V- or U-bottom wells
- Up to 10 µl of fluid per tip aspirated
- Sturdy, durable polypropylene construction
- Re-usable and steam sterilisable