Orders LinkContactUsLinkComponent
Sample containers, with screw cap
Sample containers, with screw cap
Sample containers, with screw cap
Supplier:  DELTALAB
Certificates
Sample containers, with screw cap
Sample containers, with screw cap
3 Options Available Below
Product Details & Documents
Container made of PS with PE screw cap.
  • Good chemical resistance
  • Aseptic or sterile available
  • With high safety closure biflex caps
Recommended volume: 50 ml
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Product Family Options
Product Information
  • Sekvence
    Kapacita
    Popis
    Výška
    Barva
    Barva víčka
    Ø horní části / Ø dna (mm)
    Verze
    Availability
    Price
  • 01
    60 ml
    Sample container
    66 mm
    Průhledná
    Červený
    39,3/44,2 mm
    aseptic
    Log in for availability
    Log in for availability
    1 * 600 KS
    1 * 600 KS - 6 560,00 Kč
    6 560,00 Kč
    1 * 600 KS
       6 560,00 Kč
  • 03
    60 ml
    Sample container, with spoon
    66 mm
    Průhledná
    Červený
    39,3/44,2 mm
    aseptic
    Log in for availability
    Log in for availability
    1 * 600 KS
    1 * 600 KS - 10 020,00 Kč
    10 020,00 Kč
    1 * 600 KS
       10 020,00 Kč
  • 03
    60 ml
    Sample container, individually wrapped
    66 mm
    Průhledná
    Červený
    39,3/44,2 mm
    sterile
    Log in for availability
    Log in for availability
    1 * 750 KS
    1 * 750 KS - 8 800,00 Kč
    8 800,00 Kč
    1 * 750 KS
       8 800,00 Kč