Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Catalog # 392-0371
Supplier: Greiner Bio-One
Specifications
- Doporučený pracovní objem:4 - 25 µl
- Barva:Bílá
- Popis:Solid bottom, HiBase, Low Volume™, with lid
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Plocha pro kultury:0,27 cm²
- Bal.:32
- Objem jamky:30 µl
Specifications
About this item
PS, flat bottom, round wells, TC treated. These 384-well Low Volume™ microplates offer the same standardised footprint and height as regular polystyrene plates. This enables a direct change from larger volumes into the small volume range or even a combination of both plates.
- Raised rims on each well minimise cross contamination
- Small volume allows reagent savings