Flow through cooler CK 310
Catalog # SCOI285414320
Supplier: SI Analytics
Specifications
- Popis:Flow-through cooler
- Bal.:1
- Typ:CK 310
Specifications
About this item
The flow-through cooler CK 310 is the perfect complement to bath thermostats of CT 72 series to extending their temperature range to below ambient temperature.
- Small footprint
- Low energy requirement: Economic solution preventing tap water usage for cooling
- Internal cooling loop is made of stainless steel: Better chemical resistance than copper