Viscosity flow cups
Dodavatel: Industrial Physics
Přejít na:
O tomto produktu
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certifications: AFNOR
4 Dostupných variant níže
Product Details & Documents
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certifications: AFNOR
Documents
TQC Company Video
Videos
Product Family Options
Informace o produktu
- SekvenceModelDostupnostPrice
- SIMEVF2195Viscosity flow cup
- SIMEVF2196Viscosity flow cup
- SIMEVF2197Viscosity flow cup
- SIMEVF2198Viscosity flow cup
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Temperature control jacket with tripod for ISO and AFNOR cupsTemperature control jacket with tripod for ISO and AFNOR cupsSIMEVF206802Přihlásit se pro dostupnost1 * 1 KS - 17 670,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Tripod stand, 260 mm
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Digital stopwatch, 0,01 s
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Ring stand fro flow cups
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Viscosity convertor of flow time to viscosityViscosity convertor of flow time to viscosity769-115101Přihlásit se pro dostupnost1 * 1 KS - 2 470,00 Kč