Sampling spoons, wide handle, eco-friendly, SP Bel-Art
BELAF367401025
- Kapacita:20 ml
- Sekvence:05
- Environmentally Preferable:Waste Reducing
- Bal.:10
PLA (polylactic acid) resin, eco-friendly, transparent.
- Green alternative to petroleum-based sampling tools; helps meet sustainability goals
- FDA grade spoons are safe for food, drug and cosmetic contact
- Comparable in strength and durability to PS, PP and other plastic resin sampling tools
- Perfect for retrieving small, specific volume samples of powders or liquids
- Wide handles for firm grip, provide just enough stiffness to easily sample packed powders and other firm materials
- Unique flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less non renewable energy from source to resin than its petroleum-based counterparts. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.