Manual compression sealer for AxyMat™ sealing mats, Axygen® EASY PRESS
Catalog # 732-0907
Supplier: Corning
Specifications
- Popis:Axygen® EASY PRESS, manual compression sealer for AxyMat™ sealing mats
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:I-RECs, ISO 14001 - Reynosa
- Bal.:1
Specifications
About this item
Axygen® EASY PRESS is a manual device which provides simple and effective application of Axymat™ and/or Impermamat™ sealing mats to 24, 48, 96 or 384 well microplates, deep well plates, microtitre tube systems and PCR plates. The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2 ml deep well blocks) using the large thumbwheel.