1-Allylimidazole
Catalog # ABCRMP-9525.0025
Supplier: abcr
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
:108.14 g/mol
EINECS:250-618-1
CAS Number:31410-01-2
Formula:C₆H₈N₂
MDL Number:MFCD00037894