Staining jar acc.to Coplin
Katalogové číslo 631-9730
Dodavatel: DELTALAB
Specifikace
- Ø:60 mm
- Popis:Staining jar, Coplin
- Výška:110 mm
- Bal.:1
Specifikace
O tomto produktu
Jar made of PP. Rectangular container with rounded base and screw ribbed flat cap with an inner ring for preventing leakage.
- Unbreakable
- Ideal for used as a storage system
- Good chemical and temperature resistance
These staining jar hold up to 10 slides (Two per groove).