Lead(II) oxide (Litharge), granular gold-, silver-free
Catalog # 029-930-11
Supplier: VWR Collection
CAS Number:
Specifications
- Bal.:25 kg
- Sekvence:4283
Specifications
Specification Test Results
- Appearance granularity:Granular
- Appearance colour:Yellow to orange
- Gold (Au):∼ 1 ppb
- Silver (Ag):max 0.16 ppm
Specification Test Results
About this item
:
Merck Index:13,05431
Boiling Pt:1470 °C (1013 hPa)
Melting Pt:888 °C
Density:9,5 g/cm³ (20 °C)
EINECS:215-267-0
UN:2291
ADR:6.1,III
:223.21 g/mol
:GHS08
:GHS07
:GHS09
CAS Number:1317-36-8
Formula:PbO
MDL Number:MFCD00011164