Alkali blue (C.I. 42765) indicator, Supelco®
Catalog # 1.09196.0025
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:25 g
- Sekvence:3308
Specifications
Specification Test Results
- Molar Mass:595.73 g/mol
Specification Test Results
About this item
:
:595.73 g/mol
:M-Clarity™ quality level = MQ200
EINECS:215-385-2
CAS Number:1324-76-1
Formula:C₃₇H₂₉N₃O₃S
MDL Number:MFCD00083221