Heating mantles, WHLMG3ER series
: WINKLER
For round bottomed flasks. These variable volume mantels for different sizes of flask have a crocheted outer jacket made from glass silk.
- Maximum temperature 450 °C
- Mantles have three heating zones
- Detachable plug-in connection (cable length 0,5 m)
- RCD personal safety plug with 1 m cable
Support Rings
Frequently Bought Together