Weighing paper, grade MN 226
Katalogové číslo 611-9091
Dodavatel: MACHEREY-NAGEL
Specifikace
- Velikost:90×115 mm
- Bal.:100
- Hmotnost:40 g/m²
Specifikace
O tomto produktu
This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that the weighed goods can be transferred without loss.