Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Kity pro environmentální testy
Iodine indicator, Supelco®
Iodine indicator, Supelco®
Iodine indicator, Supelco®
Catalog # 1.04764.0100
Supplier:
Merck
CAS Number:
SDS
Certificates
Expand image
Iodine indicator, Supelco®
Catalog # 1.04764.0100
Supplier:
Merck
CAS Number:
SDS
Certificates
Specifications
Bal.:
100 g
Popis:
Iodine indicator
Specifications
About this item
Product Details
Documentation
Product Details
Documentation