PCR plates, 96-well, no skirt, Axygen®
Dodavatel: Corning
Certifikáty
O tomto produktu
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from DNase, RNase and endotoxins
- AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 250 μl
Compatible with (in addition to standard thermal cycling):
Plates without skirt: Strategene MX4000, BioRad iCycler, MyiQ, iQ5
Plates without skirt, with flat top: ABI and BioRad qPCR systems
Plates without skirt, with elevated wells: Amersham MegaBACE sequencer
Specifikace
- Catalog No:
- 732-0661
- 732-1021
- 731-0643
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa