ContactUsLinkComponent
Mounting plate for pipette canisters
Mounting plate for pipette canisters
Catalog # BELC1288-11111
Supplier:  BELLCO GLASS
undefined
Mounting plate for pipette canisters
Catalog # BELC1288-11111
Supplier:  BELLCO GLASS
Specifications
  • Popis:
    Mounting plate, 90×90 mm, for use with 38 mm OD glass pipette canisters
  • Bal.:
    6
Specifications
About this item
Mounting plate for pipette canisters, made from stainless steel.
  • Stabilises pipette canisters during use
  • Can be used with 38 mm Ext.Ø glass pipette canisters