Full face mask, X-plore® 6300
Catalog # 111-0777
Supplier: Dräger
Specifications
- Type:6300 EPDM/PMMA with PMMA lens and plastic frame
- Size:U*
- Environmentally Preferable:
- Bal.:1
Specifications
About this item
The reusable X-plore® 6300 is the successor to the Panorama Nova Standard – redesigned and improved with fresh colours and an integrated bar code.
- Mask body is made of highly resistant, hypoallergenic EPDM
- Double-layer face seal with triple sealing actionfor a secure and comfortable fit on just about any shape of face
- Comfortable five-point head harness ensures that the mask can be donned easily and quickly
- Head harness is wide enough to prevent any pressure points on the wearer's head
- Scratch-proof and distortionfree PMMA Plexiglas visor with 180° wide angle guarantees a large field of vision
- Robust frame is made of extra-light plastic
- Intelligent ventilation system ensures that the visor remains fog-free
Together with the respiratory filters from Dräger's comprehensive X-plore® Rd40 filter range, the mask has proven suitable for a wide range of different applications. For longer application periods this mask can be used together with the Dräger powered air purifying respirators (e.g. X-plore® 7300 or 7500 series).
Weight: Approx. 500 g
*One size fits all
Certifications: EN 136 Class 2 (CE marking), NIOSH, AS/NZS