VWR® Buffer, MES-SDS, 20X, UltraPure Grade
Catalog # K856-500ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Typ balení:Plastic bottle
- Bal.:500 mL
- Popis:MES-SDS buffer, 20X concentrate
Specifications
About this item
Useful for high resolution of proteins on neutral pH SDS-PAGE gels.
- Ultra-pure grade
- Convenient concentrations and packaging
- Good quality, consistency and resolution