(3-((3-Cholamidopropyl)dimethylamonio)-1-propansulfonát) 0.1 mol/l in aqueous solution, Millipore®
Catalog # SIAL19899-1ML-F
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 mL
- Sekvence:2070
Specifications
About this item
:
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083