Welding face shield
Welding shield, helmet-mounted, EU version, Welding protection
Katalogové číslo CENUS903NCE
Dodavatel: CENTURION SAFETY PRODUCTS
Přejít na:
Product Details & Documents
This lightweight helmet-mounted welding shield is highly impact resistant.
- Shield is made of robust Polyamide PA6 (Nylon)
- Supplied with Polycarbonate chipping lens
- Compatible with all Centurion helmets except for the Spartan helmet
Window: 110×90 mm
Certifications: EN 175
Specifikace
- Type:Welding protection
- Popis:Welding shield, helmet-mounted, EU version