Order Entry
Czech Republic
ContactUsLinkComponent
bromfenolová modř in acetone
Flammable
Irritant
bromfenolová modř in acetone
Catalog # VWRS83905.365
Supplier:  VWR Collection
CAS Number:  
Flammable
Irritant
bromfenolová modř in acetone
Catalog # VWRS83905.365
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Typ balení:
    Plastic container
  • Bal.:
    5 L
  • Sekvence:
    5190

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:204-086-2
UN:1090
ADR:3,II
 :GHS02
 :GHS07
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875